Learn More
Echelon Biosciences Research Labs PI(4.5)P2 GRIP 10 ug
SDP
Supplier:  Echelon Biosciences Research Labs G450110UG
Encompass
Each vial of PI(4,5)P2 Grip contains lyophilized, recombinant GST-tagged PLC-1 PH domain protein. This PI(4,5)P2 binding protein can be used as a control reagent with PIP Strips, PIP Arrays, and PIP Plates. It is easily reconstituted in water or buffered solutions. Each 2.5 g vial contains enough protein for use on one PIP Strip or PIP Array. PI(4,5)P2 Grip can be detected with commercial anti-GST antibodies. This item is non returnable
Catalog No. NC0476582