Learn More
Echelon Biosciences Research Labs PI(4,5)P2 PolyPIPosomes 0.1 mL
SDP
Supplier:  Echelon Biosciences Research Labs YP045100UL
Encompass
PolyPIPosomes are polymerized liposomes that show markedly increased stability (up to 6 months) vs. conventional liposomes. A biotin tag is incorporated for easy detection of PIPn-binding proteins with streptavidin reagents. PolyPIPosomes are provided as a 1mM total lipid solution containing 5% PIP. Each order of 250 L or 1 mL of PolyPIPosomes includes 100 L of Control PolyPIPosomes. This item is non returnable
Catalog No. 50-720-835