Learn More
Echelon Biosciences Research Labs PI(4)P DiC16 1 mg
SDP
Catalog No. 50720624
Encompass
Phosphatidylinositol 4-phosphate diC16 (PI(4)P diC16) is a synthetic, purified dipalmitoyl PI(4)P.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 4-phosphate (PI(4)P) is the biosynthetic precursor to PI(4,5)P2 and has an important roles in regulating sphingomyelin and glycosphingolipid metabolism and membrane trafficking at the exit of the Golgi complex. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-624 Supplier Echelon Biosciences Research Labs Supplier No. P40161MG
Phosphatidylinositol 4-phosphate diC16 (PI(4)P diC16) is a synthetic, purified dipalmitoyl PI(4)P.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 4-phosphate (PI(4)P) is the biosynthetic precursor to PI(4,5)P2 and has an important roles in regulating sphingomyelin and glycosphingolipid metabolism and membrane trafficking at the exit of the Golgi complex. This item is non returnable