Learn More
Echelon Biosciences Research Labs PI(4)P DiC4 100 ug
SDP
Supplier:  Echelon Biosciences Research Labs P4004100UG
Encompass
Phosphatidylinositol 4-phosphate diC4 (PI(4)P diC4) is a synthetic, purified dibutanoyl PI(4)P.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 4-phosphate (PI(4)P) is the biosynthetic precursor to PI(4,5)P2 and has an important roles in regulating sphingomyelin and glycosphingolipid metabolism and membrane trafficking at the exit of the Golgi complex. This item is non returnable
Catalog No. 50-720-617