Learn More
Echelon Biosciences Research Labs PI(4)P DiC8 1 mg
SDP
Supplier:  Echelon Biosciences Research Labs P40081MG
Encompass
Phosphatidylinositol 4-phosphate diC8 (PI(4)P diC8) is a synthetic, purified dioctanoyl PI(4)P.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 4-phosphate (PI(4)P) is the biosynthetic precursor to PI(4,5)P2 and has an important roles in regulating sphingomyelin and glycosphingolipid metabolism and membrane trafficking at the exit of the Golgi complex. This item is non returnable
Catalog No. 50-720-619