Learn More
Echelon Biosciences Research Labs PI(4)P DIC8 AMMONIUM 500 ug
SDP
Catalog No. 50720623
Encompass
Phosphatidylinositol 4-phosphate diC8 (PI(4)P diC8) is a synthetic, purified dioctanoyl PI(4)P.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 4-phosphate (PI(4)P) is the biosynthetic precursor to PI(4,5)P2 and has an important roles in regulating sphingomyelin and glycosphingolipid metabolism and membrane trafficking at the exit of the Golgi complex. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-623 Supplier Echelon Biosciences Research Labs Supplier No. P4008A500UG
Phosphatidylinositol 4-phosphate diC8 (PI(4)P diC8) is a synthetic, purified dioctanoyl PI(4)P.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 4-phosphate (PI(4)P) is the biosynthetic precursor to PI(4,5)P2 and has an important roles in regulating sphingomyelin and glycosphingolipid metabolism and membrane trafficking at the exit of the Golgi complex. This item is non returnable