Learn More
Echelon Biosciences Research Labs PI(5)P DiC4 1 mg
SDP
Supplier:  Echelon Biosciences Research Labs P50041MG
Encompass
Phosphatidylinositol 5-phosphate diC4 (PI(5)P diC4) is a synthetic, purified dibutanoyl PI(5)P.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 5-phosphate (PI(5)P) is a rare lipid found in the nucleus that can bind the PHD finger of the nuclear adapter protein ING2. PI(5)P is phosphorylated to PI(4,5)P2 by Type II PIP kinases. This item is non returnable
Catalog No. 50-720-638