Learn More
Echelon Biosciences Research Labs PI(5)P DIC8 AMMONIUM SALT 500 ug
SDP
Catalog No. 50720646
Encompass
Phosphatidylinositol 5-phosphate diC8 (PI(5)P diC8) is a synthetic, purified dioctanoyl PI(5)P.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 5-phosphate (PI(5)P) is a rare lipid found in the nucleus that can bind the PHD finger of the nuclear adapter protein ING2. PI(5)P is phosphorylated to PI(4,5)P2 by Type II PIP kinases. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-646 Supplier Echelon Biosciences Research Labs Supplier No. P5008A500UG
Phosphatidylinositol 5-phosphate diC8 (PI(5)P diC8) is a synthetic, purified dioctanoyl PI(5)P.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 5-phosphate (PI(5)P) is a rare lipid found in the nucleus that can bind the PHD finger of the nuclear adapter protein ING2. PI(5)P is phosphorylated to PI(4,5)P2 by Type II PIP kinases. This item is non returnable