Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PI(5)P Lipid Kinase Substrate

Catalog No. A33596
Encompass_Preferred
Change view
Click to view available options
Quantity:
400 nmol
5000 nmol
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A33596 5000 nmol
A33593 400 nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A33596 Supplier Invitrogen™ Supplier No. A33596
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

The PI(5)P Lipid Kinase Substrate has been specifically optimized for use with Type II PIP kinases (e.g., PIP4K2A)

  • The PI(5)P Lipid Kinase Substrate has been specifically optimized for use with Type II PIP kinases (e.g., PIP4K2A)
  • The substrate consists of solubilized phosphatidylinositol 5-phosphate (PI(5)P) in an aqueous buffer
  • It is used in combination with an Adapta™ Universal Kinase Assay to help identify unique kinase inhibitors
  • Validated for use with Thermo Fisher Scientific lipid kinases
  • Available in multiple pack sizes for convenient use
  • Also available in our SelectScreen™ Kinase Profiling Service
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Phosphatidylinositol 5-phosphate (PI(5)P) in 30 mM HEPES pH 7.5 buffer.
Form Powder
Product Line Invitrogen
Quantity 5000 nmol
Substrate Phosphatidylinositol 5-phosphate (PI(5)P)
Substrate Type Kinase Substrate
Shipping Condition Dry Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.