Learn More
Medchemexpress LLC PI-55 | 1122579-42-3 | 98.9% | C13H13N5O | 1 ML
SDP
Catalog No. 5000356745
Encompass_Preferred
PI-55 is a specific cytokinin receptor inhibitor that is structurally related to 6-benzylaminopurine (BAP). It competitively inhibits BAP binding on Arabidopsis-specific receptors CRE1/AHK4 and AHK3. PI-55 also inhibits cytokinin-induced haustorium formation and increases parasite aggressiveness.
  • Specific cytokinin receptor inhibitor
  • Competitively inhibits BAP binding on Arabidopsis-specific receptors CRE1/AHK4 and AHK3
  • Inhibits cytokinin-induced haustorium formation
  • Increases parasite aggressiveness
  • Causes incomplete blocking of early haustorial structure development at high concentrations (10 μM and 100 μM)
  • Reduces the overall aggressiveness of P. ramosa when applied with BAP
Catalog No. 50-003-56745 Supplier Medchemexpress LLC Supplier No. HY14151910MM/1ML
PI-55 is a specific cytokinin receptor inhibitor that is structurally related to 6-benzylaminopurine (BAP). It competitively inhibits BAP binding on Arabidopsis-specific receptors CRE1/AHK4 and AHK3. PI-55 also inhibits cytokinin-induced haustorium formation and increases parasite aggressiveness.
  • Specific cytokinin receptor inhibitor
  • Competitively inhibits BAP binding on Arabidopsis-specific receptors CRE1/AHK4 and AHK3
  • Inhibits cytokinin-induced haustorium formation
  • Increases parasite aggressiveness
  • Causes incomplete blocking of early haustorial structure development at high concentrations (10 μM and 100 μM)
  • Reduces the overall aggressiveness of P. ramosa when applied with BAP