Learn More
Medchemexpress LLC PI-55 | 1122579-42-3 | 98.9% | C13H13N5O | 25 MG
SDP
Catalog No. 5000352637
Encompass_Preferred
PI-55 is a specific cytokinin receptor inhibitor structurally related to 6-benzylaminopurine (BAP). It competitively inhibits BAP binding on Arabidopsis-specific receptors CRE1/AHK4 and AHK3, inhibiting cytokinin-induced haustorium formation and increasing parasite aggressiveness.
  • Specific cytokinin receptor inhibitor
  • Competitively inhibits BAP binding
  • Targets Arabidopsis-specific receptors CRE1/AHK4 and AHK3
  • Inhibits cytokinin-induced haustorium formation
  • Increases parasite aggressiveness
  • Appears as an off-white to light yellow solid
Catalog No. 50-003-52637 Supplier Medchemexpress LLC Supplier No. 25MGHY141519
PI-55 is a specific cytokinin receptor inhibitor structurally related to 6-benzylaminopurine (BAP). It competitively inhibits BAP binding on Arabidopsis-specific receptors CRE1/AHK4 and AHK3, inhibiting cytokinin-induced haustorium formation and increasing parasite aggressiveness.
  • Specific cytokinin receptor inhibitor
  • Competitively inhibits BAP binding
  • Targets Arabidopsis-specific receptors CRE1/AHK4 and AHK3
  • Inhibits cytokinin-induced haustorium formation
  • Increases parasite aggressiveness
  • Appears as an off-white to light yellow solid