Learn More
TARGETMOL CHEMICALS INC PI-55 5MG
SDP
Catalog No. 502579622
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PI-55 (6-(2-hydroxy-3-methylbenzylamino)purine) is a cytokinin receptor inhibitor that exhibits structural similarity to 6-benzylaminopurine (BAP). It competitively inhibits BAP binding on the specific Arabidopsis receptors CRE1/AHK4 and AHK3. Furthermore PI-55 effectively suppresses cytokinins-induced haustorium formation and enhances parasite aggressiveness [1]. purity: 98%
Catalog No. 50-257-9622 Supplier TARGETMOL CHEMICALS INC Supplier No. T603905MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PI-55 (6-(2-hydroxy-3-methylbenzylamino)purine) is a cytokinin receptor inhibitor that exhibits structural similarity to 6-benzylaminopurine (BAP). It competitively inhibits BAP binding on the specific Arabidopsis receptors CRE1/AHK4 and AHK3. Furthermore PI-55 effectively suppresses cytokinins-induced haustorium formation and enhances parasite aggressiveness [1]. purity: 98%