Learn More
Medchemexpress LLC HY-108606 10mg Medchemexpress, PI-828 CAS:942289-87-4 Purity:>98%
SDP
Catalog No. 501968594
Encompass_Preferred
Medchemexpress, HY-108606 10mg PI-828 CAS:942289-87-4 PI-828 is a dual PI3K and casein kinase 2 (CK2) inhibitor with IC50s of 173 nM, 149 nM, and 1127 nM for p110α, CK2, and CK2α2 in lipid kinase assay, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8594 Supplier Medchemexpress LLC Supplier No. HY10860610MG
Medchemexpress, HY-108606 10mg PI-828 CAS:942289-87-4 PI-828 is a dual PI3K and casein kinase 2 (CK2) inhibitor with IC50s of 173 nM, 149 nM, and 1127 nM for p110α, CK2, and CK2α2 in lipid kinase assay, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.