Learn More
New England Biolabs, Inc. PI-PspI – 500 units
SDP
Catalog No. 50812402
Encompass
A restriction endonuclease that recognizes the sequence TGGCAAACAGCTA_TTAT^GGGTATTATGGGT.PI-PspI is obtained from a strain of E. coli which expresses the DNA polymerase from the extreme thermophile, Pyrococcus species GB-D (1). PI-PspI is a product of in vivo protein splicing that gives rise to both polymerase and endonuclease from a single polypeptide precursor.
Catalog No. 50-812-402 Supplier New England Biolabs, Inc. Supplier No. R0695S
A restriction endonuclease that recognizes the sequence TGGCAAACAGCTA_TTAT^GGGTATTATGGGT.PI-PspI is obtained from a strain of E. coli which expresses the DNA polymerase from the extreme thermophile, Pyrococcus species GB-D (1). PI-PspI is a product of in vivo protein splicing that gives rise to both polymerase and endonuclease from a single polypeptide precursor.