Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Pi-Pump Pipet Filler
SCI_ED

Catalog No. S37102 Shop All Fisherbrand Products
Change view
Click to view available options
Autoclavable:
Not Autoclavable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Autoclavable
S37102 Not Autoclavable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S37102
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For easy, one-handed control of filling and delivery from conventional pipets

  • Knurled thumbwheel is rotated to raise or lower plunger
  • Quick-release lever permits rapid emptying
  • Safe for use with toxic or corrosive chemicals

Specifications

Autoclavable Not Autoclavable
Color Blue
Material Polypropylene, Silicone Rubber
Type Pi-pump Pipet Filler
For Use With (Application) Pipets up to 2 mL
For Use With (Equipment) 2 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.