Learn More
New England Biolabs, Inc. PI-SceI – 250 units
SDP
Catalog No. 50812404
Encompass
A restriction endonuclease that recognizes the sequence ATCTATGTCGG_GTGC^GGAGAAAGAGGTAAT. The intein encoding PI-SceI is present in the VMA ATPase gene Saccharomyces cerevisiae. The gene has been modified for independent expression in E.coli using a T7 RNA polymerase expression system.
Catalog No. 50-812-404 Supplier New England Biolabs, Inc. Supplier No. R0696S
A restriction endonuclease that recognizes the sequence ATCTATGTCGG_GTGC^GGAGAAAGAGGTAAT. The intein encoding PI-SceI is present in the VMA ATPase gene Saccharomyces cerevisiae. The gene has been modified for independent expression in E.coli using a T7 RNA polymerase expression system.