Learn More
Aviva Systems Biology Corp PI15 Antibody - N-terminal region (ARP61010_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61010_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PI15. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human PI15. Alternate names: CRISP8, DKFZp686F0366, P24TI, P25TIApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-232