Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology PI3 Kinase Antibody Sampler Kit, Unconjugated, 1 Kit
SDP

Catalog No. 502051288
Encompass

The PI3 Kinase Antibody Sampler Kit offers an economical solution for studying PI3 kinase subunits in cellular samples. This kit includes a comprehensive selection of primary and secondary antibodies required for analyzing various catalytic and regulatory subunits involved in PI3K signaling pathways. It is designed to provide sufficient materials to perform two Western blot experiments for each primary antibody included in the set.

  • Includes antibodies for PI3K p110 alpha, p110 beta, p110 gamma, and Class III subunits
  • Contains phospho-specific antibodies for PI3K p85 and p55 subunits
  • Supplied with HRP-linked secondary antibody for convenient detection
  • Optimized for Western blotting and immunoprecipitation applications
  • Sufficient for two Western blot experiments per primary antibody

Catalog No. 50-205-1288 Supplier Cell Signaling Technology Supplier No. 9655T
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The PI3 Kinase Antibody Sampler Kit offers an economical solution for studying PI3 kinase subunits in cellular samples. This kit includes a comprehensive selection of primary and secondary antibodies required for analyzing various catalytic and regulatory subunits involved in PI3K signaling pathways. It is designed to provide sufficient materials to perform two Western blot experiments for each primary antibody included in the set.

  • Includes antibodies for PI3K p110 alpha, p110 beta, p110 gamma, and Class III subunits
  • Contains phospho-specific antibodies for PI3K p85 and p55 subunits
  • Supplied with HRP-linked secondary antibody for convenient detection
  • Optimized for Western blotting and immunoprecipitation applications
  • Sufficient for two Western blot experiments per primary antibody

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.