Learn More
ABclonal Technology PI3 Kinase p110 alpha
SDP
Catalog No. 5000006584
Encompass_Preferred
Phosphatidylinositol 3-kinase is composed of an 85 kDa regulatory subunit and a 110 kDa catalytic subunit The protein encoded by this gene represents the catalytic subunit which uses ATP to phosphorylate PtdIns PtdIns4P and PtdIns(4 5)P2 This gene has been found to be oncogenic and has been implicated in cervical cancers A pseudogene of this gene has been defined on chromosome 22
Catalog No. 50-000-06584 Supplier ABclonal Technology Supplier No. A2273020UL
Phosphatidylinositol 3-kinase is composed of an 85 kDa regulatory subunit and a 110 kDa catalytic subunit The protein encoded by this gene represents the catalytic subunit which uses ATP to phosphorylate PtdIns PtdIns4P and PtdIns(4 5)P2 This gene has been found to be oncogenic and has been implicated in cervical cancers A pseudogene of this gene has been defined on chromosome 22