Learn More
Medchemexpress LLC PI3-Kinase p110 β Antibody (YA5808) | Predicted band size: 123 kDa; Observed band size: 110 kDa | 50 UL
SDP
Catalog No. 5000448554
Encompass_Preferred
PI3-Kinase p110 β is a phosphoinositide-3-kinase (PI3K) pivotal for cell signaling. It phosphorylates phosphatidylinositol derivatives, producing PIP3, which activates cascades vital for cell growth, survival, proliferation, and motility. This kinase is involved in AKT1 activation, platelet function, autophagy, insulin signaling, and mediates oncogenic signals.
  • Rabbit-derived, non-conjugated IgG monoclonal antibody
  • Reacts with human, mouse, and rat samples
  • Purified by protein A
  • Supplied in PBS, 50% glycerol, 0.05% Proclin 300, 0.05%BSA
  • Suitable for IHC-P, WB, ICC/IF, ELISA, and IP applications
  • Plays crucial roles in cell growth, survival, proliferation, and motility signaling
  • Involved in AKT1 activation and platelet function
  • Impacts autophagy, insulin signaling, and clathrin-mediated endocytosis
Catalog No. 50-004-48554 Supplier Medchemexpress LLC Supplier No. HYP8611650UL
Add to Cart
Add to Cart
PI3-Kinase p110 β is a phosphoinositide-3-kinase (PI3K) pivotal for cell signaling. It phosphorylates phosphatidylinositol derivatives, producing PIP3, which activates cascades vital for cell growth, survival, proliferation, and motility. This kinase is involved in AKT1 activation, platelet function, autophagy, insulin signaling, and mediates oncogenic signals.
  • Rabbit-derived, non-conjugated IgG monoclonal antibody
  • Reacts with human, mouse, and rat samples
  • Purified by protein A
  • Supplied in PBS, 50% glycerol, 0.05% Proclin 300, 0.05%BSA
  • Suitable for IHC-P, WB, ICC/IF, ELISA, and IP applications
  • Plays crucial roles in cell growth, survival, proliferation, and motility signaling
  • Involved in AKT1 activation and platelet function
  • Impacts autophagy, insulin signaling, and clathrin-mediated endocytosis