Learn More
Eurofins Discoverx PI3 Kinase (p110α/p65α), 10 µg. (14-790)
Supplier:  Eurofins Discoverx 14790
Encompass
Complex of N-terminal 6xHis-tagged recombinant, human full length p110α and untagged, recombinant, full length Human p65α. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9349