Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PI3 Kinase p55 gamma Antibody | 50 UL
SDP

Catalog No. 5000442508
Encompass_Preferred

PI3 Kinase p55 gamma Antibody is a Rabbit-derived and non-conjugated IgG polyclonal antibody, targeting to PI3 Kinase p55 gamma. It functions by binding to activated (phosphorylated) protein-tyrosine kinases through its SH2 domain and regulates their kinase activity. During insulin stimulation, it also binds to IRS-1. This antibody is for research use only.

  • Host: Rabbit
  • Clonality: Polyclonal
  • Species reactivity: Human, Mouse, Rat
  • Sensitivity: Endogenous
  • Purity: Affinity purified
  • Conjugation: Non-conjugated
  • Isotype: IgG
  • Appearance: Liquid
  • Storage: Stored at -20°C for 1 year, avoid repeated freeze/thaw cycles
  • Shipping: With blue ice
  • Applications: WB, IHC-P, ICC/IF
  • Molecular weight: 54 kDa

Catalog No. 50-004-42508 Supplier Medchemexpress LLC Supplier No. HYP8086650UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PI3 Kinase p55 gamma Antibody is a Rabbit-derived and non-conjugated IgG polyclonal antibody, targeting to PI3 Kinase p55 gamma. It functions by binding to activated (phosphorylated) protein-tyrosine kinases through its SH2 domain and regulates their kinase activity. During insulin stimulation, it also binds to IRS-1. This antibody is for research use only.

  • Host: Rabbit
  • Clonality: Polyclonal
  • Species reactivity: Human, Mouse, Rat
  • Sensitivity: Endogenous
  • Purity: Affinity purified
  • Conjugation: Non-conjugated
  • Isotype: IgG
  • Appearance: Liquid
  • Storage: Stored at -20°C for 1 year, avoid repeated freeze/thaw cycles
  • Shipping: With blue ice
  • Applications: WB, IHC-P, ICC/IF
  • Molecular weight: 54 kDa

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.