Learn More
ABclonal Technology PI3 Kinase p85 alpha/beta
SDP
Supplier:  ABclonal Technology A22487100UL
Encompass_Preferred
Phosphatidylinositol 3-kinase phosphorylates the inositol ring of phosphatidylinositol at the 3-prime position The enzyme comprises a 110 kD catalytic subunit and a regulatory subunit of either 85 55 or 50 kD This gene encodes the 85 kD regulatory subunit Phosphatidylinositol 3-kinase plays an important role in the metabolic actions of insulin and a mutation in this gene has been associated with insulin resistance Alternative splicing of this gene results in four transcript variants encoding different isoforms [provided by RefSeq Jun 2011]
Catalog No. 50-000-06954