Learn More
Medchemexpress LLC PI3 Kinase p85 beta Antibody (YA688) | 100 UL
SDP
Catalog No. 5000441852
Encompass_Preferred
PI3 Kinase p85 beta Antibody (YA688) is a mouse-derived and non-conjugated IgG1 monoclonal antibody, targeting to PI3 Kinase p85 beta. This antibody is affinity purified and is intended for research use only. It serves as a regulatory subunit of phosphoinositide-3-kinase (PI3K), playing a key role in cell growth, survival, proliferation, motility, and morphology by activating relevant signaling cascades. The antibody also indirectly regulates autophagy and promotes nuclear translocation of XBP1 isoform 2.
  • Mouse-derived IgG1 monoclonal antibody
  • Targets PI3 Kinase p85 beta
  • Affinity purified
  • Supplied in 1*PBS (pH 7.3), 50% glycerol and 0.5% BSA
  • Contains 0.02% sodium azide as a preservative
  • Stable for 1 year when stored at -20°C; avoid repeated freeze/thaw cycles
  • Shipped with blue ice
  • Regulates phosphoinositide-3-kinase (PI3K)
  • Activates signaling cascades for cell growth, survival, and proliferation
Catalog No. 50-004-41852 Supplier Medchemexpress LLC Supplier No. HYP80868100UL
PI3 Kinase p85 beta Antibody (YA688) is a mouse-derived and non-conjugated IgG1 monoclonal antibody, targeting to PI3 Kinase p85 beta. This antibody is affinity purified and is intended for research use only. It serves as a regulatory subunit of phosphoinositide-3-kinase (PI3K), playing a key role in cell growth, survival, proliferation, motility, and morphology by activating relevant signaling cascades. The antibody also indirectly regulates autophagy and promotes nuclear translocation of XBP1 isoform 2.
  • Mouse-derived IgG1 monoclonal antibody
  • Targets PI3 Kinase p85 beta
  • Affinity purified
  • Supplied in 1*PBS (pH 7.3), 50% glycerol and 0.5% BSA
  • Contains 0.02% sodium azide as a preservative
  • Stable for 1 year when stored at -20°C; avoid repeated freeze/thaw cycles
  • Shipped with blue ice
  • Regulates phosphoinositide-3-kinase (PI3K)
  • Activates signaling cascades for cell growth, survival, and proliferation