Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pi3 kinase p85 beta antibody (YA688) | 50 UL
SDP

Catalog No. 5000442215
Encompass_Preferred

PI3 Kinase p85 beta Antibody (YA688) is a Mouse-derived and non-conjugated IgG1 monoclonal antibody, targeting to PI3 Kinase p85 beta. This antibody is intended for research use only.

  • Host: Mouse
  • Clonality: Monoclonal
  • Species reactivity: Human, Mouse, Rat
  • Immunogen: Synthetic peptide corresponding to human PI3 Kinase p85 beta
  • Application & dilution ratio: WB (Western blot) 1:500-1:1000
  • Conjugation: Non-conjugated
  • Isotype: IgG1
  • Appearance: Liquid
  • Formulation: Supplied in 1*PBS (pH 7.3), 50% glycerol and 0.5% BSA. Preservative: 0.02% sodium azide.
  • Storage & stability: Stored at -20°C for 1 year. Avoid repeated freeze / thaw cycles.
  • Function: Regulatory subunit of phosphoinositide-3-kinase (PI3K), phosphorylates PtdIns(4,5)P2 to generate phosphatidylinositol 3,4,5-trisphosphate (PIP3).

Catalog No. 50-004-42215 Supplier Medchemexpress LLC Supplier No. HYP8086850UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PI3 Kinase p85 beta Antibody (YA688) is a Mouse-derived and non-conjugated IgG1 monoclonal antibody, targeting to PI3 Kinase p85 beta. This antibody is intended for research use only.

  • Host: Mouse
  • Clonality: Monoclonal
  • Species reactivity: Human, Mouse, Rat
  • Immunogen: Synthetic peptide corresponding to human PI3 Kinase p85 beta
  • Application & dilution ratio: WB (Western blot) 1:500-1:1000
  • Conjugation: Non-conjugated
  • Isotype: IgG1
  • Appearance: Liquid
  • Formulation: Supplied in 1*PBS (pH 7.3), 50% glycerol and 0.5% BSA. Preservative: 0.02% sodium azide.
  • Storage & stability: Stored at -20°C for 1 year. Avoid repeated freeze / thaw cycles.
  • Function: Regulatory subunit of phosphoinositide-3-kinase (PI3K), phosphorylates PtdIns(4,5)P2 to generate phosphatidylinositol 3,4,5-trisphosphate (PIP3).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.