Learn More
Enzo Life Sciences PI31 polyclonal antibody (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW97100100
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to aa 241-271 of C-terminal of human PI31. Species reactivity: Human, Mouse. Applications: WB. Formulation: Liquid. In PBS containing 10mM sodium azide.
Catalog No. 50-201-1698