Learn More
Apexbio Technology LLC DiscoveryProbe™ PI3K/Akt/mTOR Compound Library, 100uL/well(10 mM solution), 96 Well Racks With Screw Cap
SDP
Catalog No. 501905607
Encompass_Preferred
A unique collection of 178 PI3K/Akt/mTOR inhibitors for PI3K/Akt/mTOR signaling pathway research. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5607 Supplier Apexbio Technology LLC Supplier No. L1034.1
A unique collection of 178 PI3K/Akt/mTOR inhibitors for PI3K/Akt/mTOR signaling pathway research. Other sizes are available. Please inqury us for quote.