Learn More
Medchemexpress LLC PI3K/Akt/CREB activator 1 (compound AE-18) | 2708177-73-3 | C19H15F4NO3 | 100 MG
SDP
Catalog No. 5000372361
Encompass_Preferred
PI3K/Akt/CREB activator 1 (compound AE-18) is a potent, orally active activator that promotes neuronal proliferation, induces differentiation, and accelerates axon-dendrite polarization. It functions by upregulating brain-derived neurotrophic factor via the PI3K/Akt/CREB pathway and is suitable for research into vascular dementia (VaD).
  • Promotes neuronal proliferation
  • Induces differentiation of Neuro-2a cells into a neuron-like morphology
  • Accelerates axon-dendrite polarization of primary hippocampal neurons
  • Upregulates brain-derived neurotrophic factor via the PI3K/Akt/CREB pathway
  • Mitigates impairment of learning and memory in chronic cerebral hypoperfusion models
  • Alleviates pathological injury in the hippocampus
  • Improves cerebral blood flow recovery
Catalog No. 50-003-72361 Supplier Medchemexpress LLC Supplier No. HY151527100MG
PI3K/Akt/CREB activator 1 (compound AE-18) is a potent, orally active activator that promotes neuronal proliferation, induces differentiation, and accelerates axon-dendrite polarization. It functions by upregulating brain-derived neurotrophic factor via the PI3K/Akt/CREB pathway and is suitable for research into vascular dementia (VaD).
  • Promotes neuronal proliferation
  • Induces differentiation of Neuro-2a cells into a neuron-like morphology
  • Accelerates axon-dendrite polarization of primary hippocampal neurons
  • Upregulates brain-derived neurotrophic factor via the PI3K/Akt/CREB pathway
  • Mitigates impairment of learning and memory in chronic cerebral hypoperfusion models
  • Alleviates pathological injury in the hippocampus
  • Improves cerebral blood flow recovery