Learn More
ABclonal Technology PI3K-gamma Rabbit mAb
SDP
Catalog No. 5000008895
Encompass_Preferred
Phosphoinositide 3-kinases (PI3Ks) phosphorylate inositol lipids and are involved in the immune response The protein encoded by this gene is a class I catalytic subunit of PI3K Like other class I catalytic subunits (p110-alpha p110-beta and p110-delta) the encoded protein binds a p85 regulatory subunit to form PI3K This gene is located in a commonly deleted segment of chromosome 7 previously identified in myeloid leukemias Several transcript variants encoding the same protein have been found for this gene
Catalog No. 50-000-08895 Supplier ABclonal Technology Supplier No. A27717100UL
Phosphoinositide 3-kinases (PI3Ks) phosphorylate inositol lipids and are involved in the immune response The protein encoded by this gene is a class I catalytic subunit of PI3K Like other class I catalytic subunits (p110-alpha p110-beta and p110-delta) the encoded protein binds a p85 regulatory subunit to form PI3K This gene is located in a commonly deleted segment of chromosome 7 previously identified in myeloid leukemias Several transcript variants encoding the same protein have been found for this gene