Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PI3Kδ/γ-IN-1 | 1980884-01-2 | C25H28N6O4S | 50 MG
SDP

Catalog No. 5000355049
Encompass_Preferred

PI3Kδ/γ-IN-1 is a potent, selective PI3K-δ/γ inhibitor developed for the research of hematological malignancies. This product is intended for research use only and not for human consumption or therapeutic use.

  • Potent, selective PI3K-δ/γ inhibitor.
  • Intended for research use only.
  • Appears as an off-white to gray solid.
  • Soluble in DMSO.
  • Store at 4°C, sealed, away from moisture and light.
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month, sealed and protected from moisture and light.

Catalog No. 50-003-55049 Supplier Medchemexpress LLC Supplier No. HY14499350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PI3Kδ/γ-IN-1 is a potent, selective PI3K-δ/γ inhibitor developed for the research of hematological malignancies. This product is intended for research use only and not for human consumption or therapeutic use.

  • Potent, selective PI3K-δ/γ inhibitor.
  • Intended for research use only.
  • Appears as an off-white to gray solid.
  • Soluble in DMSO.
  • Store at 4°C, sealed, away from moisture and light.
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month, sealed and protected from moisture and light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.