Learn More
Medchemexpress LLC HY-12068 5mg Medchemexpress, PI3K-IN-1 CAS:1349796-36-6 Purity:>98%
SDP
Catalog No. 502080949
Encompass_Preferred
Medchemexpress, HY-12068 5mg PI3K-IN-1 CAS:1349796-36-6 PI3K-IN-1 (XL-147 derivative 1) is a potent inhibitor of PI3K. PI3K-IN-1 (25 μM) blocks PI3K/Akt signaling pathways. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0949 Supplier Medchemexpress LLC Supplier No. HY120685MG
Medchemexpress, HY-12068 5mg PI3K-IN-1 CAS:1349796-36-6 PI3K-IN-1 (XL-147 derivative 1) is a potent inhibitor of PI3K. PI3K-IN-1 (25 μM) blocks PI3K/Akt signaling pathways. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.