Learn More
Medchemexpress LLC PI3K-IN-1 (XL-147 derivative 1) | 1349796-36-6 | 99.5% | 2 MG
SDP
Catalog No. 5000232743
Encompass_Preferred
PI3K-IN-1 (XL-147 derivative 1) is a small-molecule inhibitor of phosphoinositide 3-kinase (PI3K) supplied as a research analytical standard. It has reported activity blocking PI3K/Akt signaling at 25 μM and is provided in milligram-scale packages suitable for analytical use.
  • Small-molecule PI3K inhibitor for research applications.
  • Blocks PI3K/Akt signaling at 25 μM (manufacturer-reported activity).
  • Molecular formula C31H29N5O6S; molecular weight 599.66.
  • Supplied as an analytical/research standard in milligram-scale packages.
  • Suitable for biochemical assays and pathway studies.
Catalog No. 50-002-32743 Supplier Medchemexpress LLC Supplier No. HY120682MG
PI3K-IN-1 (XL-147 derivative 1) is a small-molecule inhibitor of phosphoinositide 3-kinase (PI3K) supplied as a research analytical standard. It has reported activity blocking PI3K/Akt signaling at 25 μM and is provided in milligram-scale packages suitable for analytical use.
  • Small-molecule PI3K inhibitor for research applications.
  • Blocks PI3K/Akt signaling at 25 μM (manufacturer-reported activity).
  • Molecular formula C31H29N5O6S; molecular weight 599.66.
  • Supplied as an analytical/research standard in milligram-scale packages.
  • Suitable for biochemical assays and pathway studies.