Learn More
Medchemexpress LLC PI3K-IN-18 dihydrochloride | 1188890-32-5 | 99.7% | 386.3 g/mol | C16H17Cl2N3O2S | 5 MG
SDP
Catalog No. 5000263556
Encompass_Preferred
PI3K-IN-18 dihydrochloride is a 4-morpholino-2-phenylquinazoline-derived inhibitor of PI3K p110α provided as the dihydrochloride salt for research applications. It is used to probe PI3K signaling and related mTOR pathway biology in preclinical studies and biochemical assays.
Catalog No. 50-002-63556 Supplier Medchemexpress LLC Supplier No. HY109633A5MG
PI3K-IN-18 dihydrochloride is a 4-morpholino-2-phenylquinazoline-derived inhibitor of PI3K p110α provided as the dihydrochloride salt for research applications. It is used to probe PI3K signaling and related mTOR pathway biology in preclinical studies and biochemical assays.