Learn More
Medchemexpress LLC HY-102031 5mg Medchemexpress, PI3Kδ-IN-2 CAS:1702816-75-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1020315MG
Encompass_Preferred
Medchemexpress, HY-102031 5mg PI3Kδ-IN-2 CAS:1702816-75-8 PI3Kδ-IN-2 is a potent and selective inhibitor of PI3Kδ extracted from patent WO 2015055071 A1, compound 10; has an IC50 of 6.4 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7959