Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PI3K-IN-31 | 1359956-12-9 | 99.3% | 435.43 g/mol | C19H23F2N7O3 | 5 MG
SDP

Catalog No. 5000222588
Encompass_Preferred

PI3K-IN-31 is a small-molecule PI3K inhibitor for research use, showing potent activity across class I PI3K isoforms and reported anticancer effects. Supplied as a high-purity solid or as a DMSO solution, it is intended for preclinical and signaling studies in oncology research.

  • Potent multi-isoform PI3K inhibition: PI3Kα 3.7 nM; PI3Kβ 74 nM; PI3Kγ 14.6 nM; PI3Kδ 9.9 nM.
  • High purity: 99.33%.
  • Available formats: solid (5, 10, 25, 50, 100 mg) and 10 mM solution in DMSO.
  • C hemical formula C19H23F2N7O3; molecular weight 435.43 g/mol.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-22588 Supplier Medchemexpress LLC Supplier No. HY1434035MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PI3K-IN-31 is a small-molecule PI3K inhibitor for research use, showing potent activity across class I PI3K isoforms and reported anticancer effects. Supplied as a high-purity solid or as a DMSO solution, it is intended for preclinical and signaling studies in oncology research.

  • Potent multi-isoform PI3K inhibition: PI3Kα 3.7 nM; PI3Kβ 74 nM; PI3Kγ 14.6 nM; PI3Kδ 9.9 nM.
  • High purity: 99.33%.
  • Available formats: solid (5, 10, 25, 50, 100 mg) and 10 mM solution in DMSO.
  • C hemical formula C19H23F2N7O3; molecular weight 435.43 g/mol.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.