Learn More
Medchemexpress LLC PI3K-IN-31 | 1359956-12-9 | 99.3% | C19H23F2N7O3 | 100 MG
SDP
Catalog No. 5000355543
Encompass_Preferred
PI3K-IN-31 (Compound 6b) is a potent PI3K inhibitor that has demonstrated anticancer effects. It selectively inhibits PI3Kα, PI3Kβ, PI3Kγ, and PI3Kδ with varying IC50 values.
  • Potent PI3K inhibitor
  • Exhibits anticancer effects
  • Selectively inhibits PI3Kα, PI3Kβ, PI3Kγ, and PI3Kδ
Catalog No. 50-003-55543 Supplier Medchemexpress LLC Supplier No. HY143403100MG
PI3K-IN-31 (Compound 6b) is a potent PI3K inhibitor that has demonstrated anticancer effects. It selectively inhibits PI3Kα, PI3Kβ, PI3Kγ, and PI3Kδ with varying IC50 values.
  • Potent PI3K inhibitor
  • Exhibits anticancer effects
  • Selectively inhibits PI3Kα, PI3Kβ, PI3Kγ, and PI3Kδ