Learn More
Medchemexpress LLC PI3K-IN-31 | 1359956-12-9 | 99.3% | C19H23F2N7O3 | 25 MG
SDP
Catalog No. 5000356973
Encompass_Preferred
PI3K-IN-31 (Compound 6b) is a potent PI3K inhibitor, demonstrating significant activity against various PI3K isoforms including PI3Kα, PI3Kβ, PI3Kγ, and PI3Kδ. This compound has been noted for its anticancer properties.
  • Potent PI3K inhibitor
  • Exhibits anticancer effects
  • Inhibits PI3Kα with an IC50 of 3.7 nM
  • Inhibits PI3Kβ with an IC50 of 74 nM
  • Inhibits PI3Kγ with an IC50 of 14.6 nM
  • Inhibits PI3Kδ with an IC50 of 9.9 nM
Catalog No. 50-003-56973 Supplier Medchemexpress LLC Supplier No. 25MGHY143403
PI3K-IN-31 (Compound 6b) is a potent PI3K inhibitor, demonstrating significant activity against various PI3K isoforms including PI3Kα, PI3Kβ, PI3Kγ, and PI3Kδ. This compound has been noted for its anticancer properties.
  • Potent PI3K inhibitor
  • Exhibits anticancer effects
  • Inhibits PI3Kα with an IC50 of 3.7 nM
  • Inhibits PI3Kβ with an IC50 of 74 nM
  • Inhibits PI3Kγ with an IC50 of 14.6 nM
  • Inhibits PI3Kδ with an IC50 of 9.9 nM