Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PI3K-IN-37 | 1257547-40-2 | 99.0% | 442.51 | 1 MG
SDP

Supplier:  Medchemexpress LLC HY1472841MG

Encompass_Preferred

PI3K-IN-37 is an inhibitor targeting PI3K α/β/δ and mTOR. It demonstrates potent inhibition with IC50 values of 6 nM for PI3Kα, 8 nM for PI3Kβ, 4 nM for PI3Kδ, and 4 nM for mTOR. This compound is a light yellow to light brown solid with a molecular formula of C25H26N6O2 and a purity of 99.0%.

  • Inhibits PI3K α, β, and δ with IC50s of 6 nM, 8 nM, and 4 nM, respectively
  • Inhibits mTOR with an IC50 of 4 nM
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month
  • For research use only

Catalog No. 50-003-62930


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.