Learn More
Medchemexpress LLC HY-131345 5mg Medchemexpress, PI3Kα-IN-4 CAS:2322293-83-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1313455MG
Encompass_Preferred
Medchemexpress, HY-131345 5mg PI3Kα-IN-4 CAS:2322293-83-2 PI3Kα-IN-4 is a potent, selective and orally active inhibitor of PI3Kα, with an IC50 of 1.8 nM. PI3Kα-IN-4 has antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1279