Learn More
Medchemexpress LLC PI3Kγ inhibitor AZ2 | 2231760-33-9 | 99.25% | 369.48 | 1 ML
SDP
Catalog No. 5000439644
Encompass_Preferred
PI3Kγ inhibitor AZ2 is a highly selective PI3Kγ inhibitor for research into inflammatory and immune diseases.
  • Highly selective PI3Kγ inhibitor (pIC50 = 9.3 for PI3Kγ)
  • Suitable for research on inflammatory and immune diseases
  • Excellent selectivity for PI3Kγ confirmed in SKOV-3 cells (0.1-100 nM; 1 hour) with a constitutively activated PI3K/Akt pathway
Catalog No. 50-004-39644 Supplier Medchemexpress LLC Supplier No. HY11157010MM/1ML
PI3Kγ inhibitor AZ2 is a highly selective PI3Kγ inhibitor for research into inflammatory and immune diseases.
  • Highly selective PI3Kγ inhibitor (pIC50 = 9.3 for PI3Kγ)
  • Suitable for research on inflammatory and immune diseases
  • Excellent selectivity for PI3Kγ confirmed in SKOV-3 cells (0.1-100 nM; 1 hour) with a constitutively activated PI3K/Akt pathway