Learn More
Medchemexpress LLC PI3K/mTOR Inhibitor-11 | 2845104-25-6 | C27H21N7 | 50 MG
SDP
Catalog No. 5000376946
Encompass_Preferred
PI3K/mTOR Inhibitor-11 is an orally active inhibitor targeting PI3Kα, PI3Kδ, and mTOR. It functions by regulating the PI3K/AKT/mTOR signaling pathway, specifically inhibiting the phosphorylation of AKT and S6 proteins, and can be applied in cancer research.
  • Orally active.
  • Inhibits key proteins in the PI3K/AKT/mTOR pathway.
  • Regulates signaling pathway.
  • Inhibits phosphorylation of AKT and S6 proteins.
  • Demonstrates antiproliferative activity against human cancer cell lines.
  • Suppresses tumor growth in xenograft models.
  • Shows good oral bioavailability.
Catalog No. 50-003-76946 Supplier Medchemexpress LLC Supplier No. HY15162250MG
PI3K/mTOR Inhibitor-11 is an orally active inhibitor targeting PI3Kα, PI3Kδ, and mTOR. It functions by regulating the PI3K/AKT/mTOR signaling pathway, specifically inhibiting the phosphorylation of AKT and S6 proteins, and can be applied in cancer research.
  • Orally active.
  • Inhibits key proteins in the PI3K/AKT/mTOR pathway.
  • Regulates signaling pathway.
  • Inhibits phosphorylation of AKT and S6 proteins.
  • Demonstrates antiproliferative activity against human cancer cell lines.
  • Suppresses tumor growth in xenograft models.
  • Shows good oral bioavailability.