Learn More
Medchemexpress LLC PI3K/mTOR inhibitor-13 sodium | 2361009-23-4 | 98.1% | 464.40 g/mol | C20H13F2N5NaO3S | 10 MG
SDP
Catalog No. 5000286541
Encompass_Preferred
PI3K/mTOR inhibitor-13 sodium is an orally active small-molecule dual inhibitor of phosphoinositide 3-kinase (PI3K) and mTOR kinases provided as the sodium salt for preclinical research applications. It is used to probe PI3K/mTOR signaling in oncology and fibrotic disease models and is supplied in small research quantities with documented purity and supporting documentation.
  • Oral dual PI3K and mTOR kinase inhibitor.
  • High reported purity (98.13%).
  • Available in small research quantities, including a 10 mg vial.
  • Suitable for in vitro and in vivo preclinical studies.
  • Includes supporting documentation such as COA and SDS.
Catalog No. 50-002-86541 Supplier Medchemexpress LLC Supplier No. HY153120A10MG
PI3K/mTOR inhibitor-13 sodium is an orally active small-molecule dual inhibitor of phosphoinositide 3-kinase (PI3K) and mTOR kinases provided as the sodium salt for preclinical research applications. It is used to probe PI3K/mTOR signaling in oncology and fibrotic disease models and is supplied in small research quantities with documented purity and supporting documentation.
  • Oral dual PI3K and mTOR kinase inhibitor.
  • High reported purity (98.13%).
  • Available in small research quantities, including a 10 mg vial.
  • Suitable for in vitro and in vivo preclinical studies.
  • Includes supporting documentation such as COA and SDS.