Learn More
Medchemexpress LLC HY-111508 10mg Medchemexpress, PI3K/mTOR Inhibitor-2 CAS:1848242-58-9 Purity:>98%
SDP
Catalog No. 501968897
Encompass_Preferred
Medchemexpress, HY-111508 10mg PI3K/mTOR Inhibitor-2 CAS:1848242-58-9 PI3K/mTOR Inhibitor-2 is a potent dual pan-PI3K/mTOR inhibitor with IC50s of 3.4/34/16/1 nM for PI3Kα/PI3Kβ/PI3Kδ/PI3Kγ and 4.7 nM for mTOR. Antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8897 Supplier Medchemexpress LLC Supplier No. HY11150810MG
Medchemexpress, HY-111508 10mg PI3K/mTOR Inhibitor-2 CAS:1848242-58-9 PI3K/mTOR Inhibitor-2 is a potent dual pan-PI3K/mTOR inhibitor with IC50s of 3.4/34/16/1 nM for PI3Kα/PI3Kβ/PI3Kδ/PI3Kγ and 4.7 nM for mTOR. Antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.