Learn More
Medchemexpress LLC PI3K/mTOR Inhibitor-2 | 1848242-58-9 | 99.1% | 478.86 | 100 MG
SDP
Catalog No. 5000345998
Encompass_Preferred
PI3K/mTOR Inhibitor-2 is a potent dual pan-PI3K/mTOR inhibitor with IC50s of 3.4/34/16/1 nM for PI3Kα/PI3Kβ/PI3Kδ/PI3Kγ and 4.7 nM for mTOR. It also exhibits antitumor activity. In MCF-7 cells, it inhibits p-AKT and p-p70s6k with IC50s of 11.6 and 89.2 nM, respectively. This compound is for research use only and has a solid appearance with a light yellow to yellow color.
  • Potent dual pan-PI3K/mTOR inhibitor
  • Exhibits antitumor activity
  • Inhibits p-AKT and p-p70s6k in MCF-7 cells
  • For research use only
  • Solid appearance, light yellow to yellow color
  • Molecular formula C20H13ClF2N4O4S
Catalog No. 50-003-45998 Supplier Medchemexpress LLC Supplier No. HY111508100MG
PI3K/mTOR Inhibitor-2 is a potent dual pan-PI3K/mTOR inhibitor with IC50s of 3.4/34/16/1 nM for PI3Kα/PI3Kβ/PI3Kδ/PI3Kγ and 4.7 nM for mTOR. It also exhibits antitumor activity. In MCF-7 cells, it inhibits p-AKT and p-p70s6k with IC50s of 11.6 and 89.2 nM, respectively. This compound is for research use only and has a solid appearance with a light yellow to yellow color.
  • Potent dual pan-PI3K/mTOR inhibitor
  • Exhibits antitumor activity
  • Inhibits p-AKT and p-p70s6k in MCF-7 cells
  • For research use only
  • Solid appearance, light yellow to yellow color
  • Molecular formula C20H13ClF2N4O4S