Learn More
Medchemexpress LLC PI3K/mTOR inhibitor-2 | 1848242-58-9 | MFCD31813651 | 99.1% | 478.86 g·mol⁻¹ | C20H13ClF2N4O4S | 5 MG
SDP
Catalog No. 5000263522
Encompass_Preferred
PI3K/mTOR Inhibitor-2 is a small-molecule research reagent that potently inhibits class I PI3K isoforms and mTOR. It shows low-nanomolar biochemical activity and is supplied for laboratory research in solid form and as a DMSO solution.
  • Potent dual inhibition of class I PI3K isoforms and mTOR (IC50s: PI3Kα 3.4 nM; PI3Kβ 34 nM; PI3Kδ 16 nM; PI3Kγ 1 nM; mTOR 4.7 nM).
  • Suitable for biochemical and cellular studies to probe PI3K/mTOR signaling.
  • High purity for research applications (reported purity 99.1%).
  • Available as a solid and as a DMSO solution for flexible dosing.
  • Molecular formula C20H13ClF2N4O4S and molecular weight 478.86 g·mol⁻¹.
Catalog No. 50-002-63522 Supplier Medchemexpress LLC Supplier No. HY1115085MG
PI3K/mTOR Inhibitor-2 is a small-molecule research reagent that potently inhibits class I PI3K isoforms and mTOR. It shows low-nanomolar biochemical activity and is supplied for laboratory research in solid form and as a DMSO solution.
  • Potent dual inhibition of class I PI3K isoforms and mTOR (IC50s: PI3Kα 3.4 nM; PI3Kβ 34 nM; PI3Kδ 16 nM; PI3Kγ 1 nM; mTOR 4.7 nM).
  • Suitable for biochemical and cellular studies to probe PI3K/mTOR signaling.
  • High purity for research applications (reported purity 99.1%).
  • Available as a solid and as a DMSO solution for flexible dosing.
  • Molecular formula C20H13ClF2N4O4S and molecular weight 478.86 g·mol⁻¹.