Learn More
Sino Biological Recombinant Human PDGF alpha receptor/PDGFRA Protein (G680R, GST Tag), Active
Catalog No. 502573880
Encompass_Preferred
Recombinant human PDGFRα (G680R) (550-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-3880 Supplier Sino Biological Supplier No. P1212DG500
Recombinant human PDGFRα (G680R) (550-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.