Learn More
Sino Biological Recombinant Human PIK3C3 Protein (Full Length, GST Tag), Active
Catalog No. 502573808
Encompass_Preferred
Recombinant full length human PIK3C3 (Vps34) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-3808 Supplier Sino Biological Supplier No. P26102G500
Recombinant full length human PIK3C3 (Vps34) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.