Learn More
Sino Biological Recombinant Human PI3K (p110 alpha/p85 alpha) Protein (p110 alpha (N345K)/ p85 alpha, His Tag), Active
Catalog No. 502573798
Encompass_Preferred
Recombinant full length human p110 alpha (N345K) mutant subunit and human p85 alpha wild-type subunit were co-expressed by baculovirus in Sf9 insect cells using an N-terminal His tag on both proteins.
Catalog No. 50-257-3798 Supplier Sino Biological Supplier No. P27122CH500
Recombinant full length human p110 alpha (N345K) mutant subunit and human p85 alpha wild-type subunit were co-expressed by baculovirus in Sf9 insect cells using an N-terminal His tag on both proteins.