Learn More
Eurofins Discoverx PI3 Kinase (p110α/p65α) murine, 10 µg. (14-786)
Supplier:  Eurofins Discoverx 14786
Encompass
Complex of N-terminal 6xHis-tagged recombinant, murine full length p110α and untagged, recombinant, full length murine p65α. p65α is an oncogenic mutant form of p85α(residues 1-571) which leads to increased PI3K activity. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9341