Learn More
Eurofins Discoverx PI3-Kinase (p110δ/p85α) murine, 250 µg. (14-789M)
Supplier:  Eurofins Discoverx 14789M
Encompass
Complex of N-terminal 6xHis-tagged recombinant, murine p110δ full length and untagged, recombinant, murine p85α full length. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9348